Assay Details
Target Gene Details
Entrez Gene ID: | 343641 |
Gene Name: | transglutaminase 6 |
Gene Aliases: |
SCA35, TG6, TGM3L, TGY, dJ734P14.3 |
Location: |
Chr.20:2380908-2432753 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGM6 | NM_001254734.1 | NP_001241663.1 | ||
| NM_198994.2 | NP_945345.2 | |||
| AF540969.1 | AAQ10751.1 | |||
| AF540970.1 | AAQ10752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|
| esv3644998 | Chr.20:2148722 - 2690694 on Build GRCh38 | Gain |
|
| esv2758778 | Chr.20:2149914 - 2494823 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map