Assay Details
Target Gene Details
Entrez Gene ID: | 245932 |
Gene Name: | defensin beta 119 |
Gene Aliases: |
DEFB-19, DEFB-20, DEFB120, DEFB20, ESC42-RELA, ESC42-RELB |
Location: |
Chr.20:31377163-31390649 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DEFB119 | NM_153289.3 | NP_695021.2 | ||
| NR_073151.1 | ||||
| NR_073152.1 | ||||
| NR_073153.1 | ||||
| NR_126440.1 | ||||
| AF479698.1 | AAQ05837.1 | |||
| AL732301.1 | ||||
| AW003815.1 | ||||
| AY122472.1 | AAM93914.1 | |||
| AY358860.1 | AAQ89219.1 | |||
| AY501001.1 | AAS87295.1 | |||
| BC062212.1 | AAH62212.1 | |||
| BI460707.1 | ||||
| BU561156.1 | ||||
| HY035342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470546 | Chr.20:31245806 - 31647499 on Build GRCh38 | Gain |
|
| nsv1060041 | Chr.20:31339025 - 31428640 on Build GRCh38 | Gain |
|
| esv2763228 | Chr.20:31247246 - 31651719 on Build GRCh38 | Gain |
|
| dgv4299n100 | Chr.20:31247234 - 31436365 on Build GRCh38 | Gain |
|
| esv3645647 | Chr.20:31367312 - 31405042 on Build GRCh38 | Gain |
|
| nsv585910 | Chr.20:31305698 - 31457590 on Build GRCh38 | Gain |
|
| esv3645644 | Chr.20:31257358 - 31413171 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114442976] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map