Assay Details
Target Gene Details
Entrez Gene ID: | 54453 |
Gene Name: | Ras and Rab interactor 2 |
Gene Aliases: |
MACS, RASSF4 |
Location: |
Chr.20:19757610-20002459 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIN2 | XM_005260731.2 | XP_005260788.1 | ||
| XM_006723574.3 | XP_006723637.1 | |||
| XM_006723575.3 | XP_006723638.1 | |||
| XM_017027887.1 | XP_016883376.1 | |||
| XM_017027888.1 | XP_016883377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645460 | Chr.20:19760368 - 19768597 on Build GRCh38 | Loss |
|
| esv3645457 | Chr.20:19637825 - 19995319 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map