Assay Details
Target Gene Details
Entrez Gene ID: | 1137 |
Gene Name: | cholinergic receptor nicotinic alpha 4 subunit |
Gene Aliases: |
BFNC, EBN, EBN1, NACHR, NACHRA4, NACRA4 |
Location: |
Chr.20:63343310-63375392 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA4 | XM_017027624.1 | XP_016883113.1 | ||
| XM_017027625.1 | XP_016883114.1 |
Target Gene Details
Entrez Gene ID: | 100130587 |
Gene Name: | uncharacterized LOC100130587 |
Gene Aliases: |
- |
Location: |
Chr.20:63359988-63371177 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130587 | NR_110634.1 | |||
| AK127527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953306 | Chr.20:63151949 - 63571247 on Build GRCh38 | Deletion |
|
| nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map