Assay Details
Target Gene Details
Entrez Gene ID: | 81030 |
Gene Name: | Z-DNA binding protein 1 |
Gene Aliases: |
C20orf183, DAI, DLM-1, DLM1 |
Location: |
Chr.20:57603846-57620576 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBP1 | NM_001160417.1 | NP_001153889.1 | ||
| NM_001160418.1 | NP_001153890.1 | |||
| NM_001160419.2 | NP_001153891.1 | |||
| NM_001323966.1 | NP_001310895.1 | |||
| NM_030776.2 | NP_110403.2 | |||
| NR_136660.1 | ||||
| XM_011529057.2 | XP_011527359.1 | |||
| XM_011529058.2 | XP_011527360.1 | |||
| XM_017028086.1 | XP_016883575.1 | |||
| AJ300575.1 | CAC18810.1 | |||
| AK122761.1 | ||||
| AK128405.1 | 1 | 1150 | ||
| AK290918.1 | ||||
| AK314062.1 | ||||
| AX775973.1 | CAE11686.1 | |||
| AX775975.1 | CAE11687.1 | |||
| BC028218.1 | AAH28218.1 | |||
| BC131706.1 | ||||
| CT002036.1 | ||||
| DA674238.1 | ||||
| DA676308.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758802 | Chr.20:57595722 - 58113215 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map