Assay Details
Target Gene Details
Entrez Gene ID: | 128486 |
Gene Name: | fat storage inducing transmembrane protein 2 |
Gene Aliases: |
C20orf142, Fit2, dJ881L22.2 |
Location: |
Chr.20:44306557-44311249 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FITM2 | NM_001080472.2 | 2 | 401 | NP_001073941.1 |
| BC130446.1 | 2 | 330 | ||
| BC136640.1 | 2 | 330 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| nsv586065 | Chr.20:44261867 - 44505244 on Build GRCh38 | Gain |
|
| nsv3389 | Chr.20:44304429 - 44338914 on Build GRCh38 | Insertion |
|
| esv3645906 | Chr.20:44266079 - 44338614 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map