Assay Details
Target Gene Details
Entrez Gene ID: | 7165 |
Gene Name: | tumor protein D52 like 2 |
Gene Aliases: |
D54, TPD54 |
Location: |
Chr.20:63865228-63891545 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509784 | Chr.20:63824382 - 63983325 on Build GRCh38 | Insertion |
|
| nsv428381 | Chr.20:63583375 - 64334167 on Build GRCh38 | Gain+Loss |
|
| nsv469773 | Chr.20:63845428 - 64039312 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map