Assay Details
Target Gene Details
Entrez Gene ID: | 80343 |
Gene Name: | SEL1L2 ERAD E3 ligase adaptor subunit |
Gene Aliases: |
C20orf50 |
Location: |
Chr.20:13849188-13995333 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEL1L2 | NM_001271539.1 | NP_001258468.1 | ||
| NR_073206.1 | ||||
| NR_073207.1 | ||||
| XM_006723646.2 | XP_006723709.1 | |||
| XM_006723647.2 | XP_006723710.1 | |||
| XM_006723648.2 | XP_006723711.1 | |||
| XM_006723650.2 | XP_006723713.1 | |||
| XM_006723651.2 | XP_006723714.1 | |||
| XM_006723652.2 | XP_006723715.1 | |||
| XM_006723654.3 | XP_006723717.1 | |||
| XM_011529374.1 | XP_011527676.1 | |||
| XM_011529375.1 | XP_011527677.1 | |||
| XM_011529376.1 | XP_011527678.1 | |||
| XM_011529377.1 | XP_011527679.1 | |||
| XM_011529378.1 | XP_011527680.1 | |||
| XM_011529379.1 | XP_011527681.1 | |||
| XM_017028084.1 | XP_016883573.1 | |||
| AK302170.1 | ||||
| AL137678.1 | ||||
| DA857535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3295 | Chr.20:13824349 - 13859019 on Build GRCh38 | Insertion |
|
| esv3645294 | Chr.20:13849940 - 13862262 on Build GRCh38 | Gain |
|

Genomic Map