Assay Details
Target Gene Details
Entrez Gene ID: | 9885 |
Gene Name: | oxysterol binding protein like 2 |
Gene Aliases: |
DFNA67, DNFA67, ORP-2, ORP2 |
Location: |
Chr.20:62238485-62296213 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OSBPL2 | NM_001278649.1 | NP_001265578.1 | ||
| NM_014835.3 | NP_055650.1 | |||
| NM_144498.2 | NP_653081.1 | |||
| XM_017028163.1 | XP_016883652.1 | |||
| XM_017028164.1 | XP_016883653.1 | |||
| XM_017028165.1 | XP_016883654.1 | |||
| XM_017028166.1 | XP_016883655.1 | |||
| XM_017028167.1 | XP_016883656.1 | |||
| XM_017028168.1 | XP_016883657.1 | |||
| XM_017028169.1 | XP_016883658.1 | |||
| XM_017028170.1 | XP_016883659.1 | |||
| AB018315.1 | ||||
| AF392447.1 | AAL40660.1 | |||
| AK000230.1 | ||||
| AK291851.1 | ||||
| AK296595.1 | ||||
| AK315576.1 | ||||
| AY028168.1 | AAK18044.1 | |||
| BC000296.2 | AAH00296.1 | |||
| BC004455.1 | AAH04455.1 | |||
| DA301911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422398 | Chr.20:62207833 - 62674040 on Build GRCh38 | Duplication |
|
| nsv1160689 | Chr.20:62188341 - 62304172 on Build GRCh38 | Deletion |
|
| nsv518296 | Chr.20:62203245 - 62415019 on Build GRCh38 | Loss |
|
| nsv470556 | Chr.20:62212004 - 62577283 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map