Assay Details
Target Gene Details
Entrez Gene ID: | 1137 |
Gene Name: | cholinergic receptor nicotinic alpha 4 subunit |
Gene Aliases: |
BFNC, EBN, EBN1, NACHR, NACHRA4, NACRA4 |
Location: |
Chr.20:63343310-63375392 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA4 | NM_000744.6 | NP_000735.1 | ||
| NM_001256573.1 | NP_001243502.1 | |||
| NR_046317.1 | ||||
| XM_011528524.1 | XP_011526826.1 | |||
| XM_017027624.1 | XP_016883113.1 | |||
| XM_017027625.1 | XP_016883114.1 | |||
| AK314907.1 | ||||
| BC096290.1 | AAH96290.1 | |||
| BC096291.1 | AAH96291.1 | |||
| BC096292.1 | AAH96292.1 | |||
| BC096293.3 | AAH96293.1 | |||
| L35901.1 | AAA64743.1 | |||
| U62433.1 | AAB40111.1 | |||
| X87629.1 | CAA60959.1 | |||
| X89743.1 | ||||
| Y08421.1 | CAA69698.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953306 | Chr.20:63151949 - 63571247 on Build GRCh38 | Deletion |
|
| nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map