Assay Details
Target Gene Details
Entrez Gene ID: | 100131208 |
Gene Name: | SNAP25 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:10023812-10219506 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNAP25-AS1 | NR_040710.1 | |||
| AK124961.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833918 | Chr.20:9950381 - 10077106 on Build GRCh38 | Loss |
|
| nsv522874 | Chr.20:9918791 - 10838562 on Build GRCh38 | Loss |
|
| nsv585414 | Chr.20:10005758 - 10050283 on Build GRCh38 | Loss |
|
| nsv1063924 | Chr.20:10003340 - 10100148 on Build GRCh38 | Gain |
|

Genomic Map