Assay Details
Target Gene Details
Entrez Gene ID: | 10326 |
Gene Name: | signal regulatory protein beta 1 |
Gene Aliases: |
CD172b, SIRP-BETA-1 |
Location: |
Chr.20:1564193-1620457 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRPB1 | XM_005260641.3 | XP_005260698.1 | ||
| XM_005260643.3 | XP_005260700.1 | |||
| XM_005260644.3 | XP_005260701.1 | |||
| XM_017027577.1 | XP_016883066.1 | |||
| XM_017027578.1 | XP_016883067.1 | |||
| XM_017027579.1 | XP_016883068.1 | |||
| XM_017027580.1 | XP_016883069.1 | |||
| XM_017027581.1 | XP_016883070.1 | |||
| XM_017027582.1 | XP_016883071.1 | |||
| XM_017027583.1 | XP_016883072.1 | |||
| XM_017027584.1 | XP_016883073.1 | |||
| XM_017027585.1 | XP_016883074.1 | |||
| AK313517.1 | ||||
| BC025286.1 | AAH25286.1 | |||
| BC075835.1 | AAH75835.1 | |||
| BC131747.1 | ||||
| BM010247.1 | ||||
| DA331023.1 | ||||
| Y10376.1 | CAA71404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585206 | Chr.20:1405829 - 1733716 on Build GRCh38 | Gain |
|
| esv2722071 | Chr.20:1565136 - 1694217 on Build GRCh38 | Deletion |
|
| nsv833894 | Chr.20:1538787 - 1772720 on Build GRCh38 | Gain |
|
| esv3644980 | Chr.20:1509522 - 1569123 on Build GRCh38 | Gain |
|
| nsv9775 | Chr.20:1554339 - 1583014 on Build GRCh38 | Loss |
|
| nsv3253 | Chr.20:1566446 - 1623186 on Build GRCh38 | Deletion |
|
| nsv522008 | Chr.20:1359746 - 1657932 on Build GRCh38 | Gain |
|
| nsv437183 | Chr.20:1565804 - 1588917 on Build GRCh38 | Loss |
|
| nsv1160672 | Chr.20:1564764 - 1580934 on Build GRCh38 | Deletion |
|
| nsv510779 | Chr.20:1528684 - 1633316 on Build GRCh38 | Deletion |
|
| esv2758777 | Chr.20:1496135 - 1698610 on Build GRCh38 | Gain+Loss |
|
| nsv1059730 | Chr.20:1566104 - 1581009 on Build GRCh38 | Loss |
|
| esv33848 | Chr.20:1565055 - 1570417 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map