Assay Details
Target Gene Details
Entrez Gene ID: | 3785 |
Gene Name: | potassium voltage-gated channel subfamily Q member 2 |
Gene Aliases: |
BFNC, EBN, EBN1, ENB1, HNSPC, KCNA11, KV7.2 |
Location: |
Chr.20:63400208-63472677 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ2 | NM_004518.5 | NP_004509.2 | ||
| NM_172106.2 | NP_742104.1 | |||
| NM_172107.3 | NP_742105.1 | |||
| NM_172108.4 | NP_742106.1 | |||
| XM_011528810.1 | XP_011527112.1 | |||
| XM_011528811.1 | XP_011527113.1 | |||
| XM_017027841.1 | XP_016883330.1 | |||
| XM_017027842.1 | XP_016883331.1 | |||
| XM_017027843.1 | XP_016883332.1 | |||
| XM_017027844.1 | XP_016883333.1 | |||
| XM_017027845.1 | XP_016883334.1 | |||
| AF033348.1 | AAB97315.1 | |||
| AF074247.1 | AAC25921.1 | |||
| AF110020.1 | AAD16988.1 | |||
| Y15065.1 | CAA75348.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953306 | Chr.20:63151949 - 63571247 on Build GRCh38 | Deletion |
|
| esv3646317 | Chr.20:63411810 - 63431038 on Build GRCh38 | Gain |
|
| esv3568097 | Chr.20:63413305 - 63423602 on Build GRCh38 | Loss |
|
| nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain |
|
| nsv470562 | Chr.20:63362590 - 63460989 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map