Assay Details
Target Gene Details
Entrez Gene ID: | 359710 |
Gene Name: | BPI fold containing family B member 3 |
Gene Aliases: |
C20orf185, LPLUNC3, RYA3 |
Location: |
Chr.20:33055331-33073637 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPIFB3 | NM_182658.2 | NP_872599.1 | ||
| AF549189.1 | AAP84989.1 | |||
| BC113398.1 | ||||
| BC113400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
| esv2758792 | Chr.20:32908001 - 33160360 on Build GRCh38 | Loss |
|
| nsv1062857 | Chr.20:33032555 - 33066287 on Build GRCh38 | Gain |
|
| nsv3352 | Chr.20:33022133 - 33066950 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map