Assay Details
Target Gene Details
Entrez Gene ID: | 284759 |
Gene Name: | signal regulatory protein beta 2 |
Gene Aliases: |
PTPN1L, PTPNS1L3, dJ776F14.2 |
Location: |
Chr.20:1474591-1491762 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRPB2 | NM_001122962.1 | NP_001116434.1 | ||
| NM_001134836.1 | NP_001128308.1 | |||
| XM_005260709.3 | XP_005260766.1 | |||
| XM_011529223.2 | 1 | 232 | XP_011527525.1 | |
| BM285394.1 | ||||
| BQ635335.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522008 | Chr.20:1359746 - 1657932 on Build GRCh38 | Gain |
|
| nsv833893 | Chr.20:1433971 - 1547133 on Build GRCh38 | Loss |
|
| nsv585206 | Chr.20:1405829 - 1733716 on Build GRCh38 | Gain |
|
| nsv952971 | Chr.20:1483656 - 1497554 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map