Assay Details
Target Gene Details
Entrez Gene ID: | 55423 |
Gene Name: | signal regulatory protein gamma |
Gene Aliases: |
CD172g, SIRP-B2, SIRPB2, SIRPgamma, bA77C3.1 |
Location: |
Chr.20:1629152-1686516 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRPG | XM_011529286.2 | XP_011527588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758777 | Chr.20:1496135 - 1698610 on Build GRCh38 | Gain+Loss |
|
| esv3644988 | Chr.20:1626418 - 1861080 on Build GRCh38 | Gain |
|
| nsv585206 | Chr.20:1405829 - 1733716 on Build GRCh38 | Gain |
|
| esv2722071 | Chr.20:1565136 - 1694217 on Build GRCh38 | Deletion |
|
| nsv833894 | Chr.20:1538787 - 1772720 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs67599103] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map