Assay Details
Target Gene Details
Entrez Gene ID: | 388795 |
Gene Name: | EF-hand calcium binding domain 8 |
Gene Aliases: |
- |
Location: |
Chr.20:32860995-32961845 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB8 | XM_011529121.1 | XP_011527423.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
| nsv833959 | Chr.20:32829604 - 32982934 on Build GRCh38 | Loss |
|
| esv996804 | Chr.20:32885915 - 32897898 on Build GRCh38 | Deletion |
|
| nsv585918 | Chr.20:32851403 - 32956284 on Build GRCh38 | Loss |
|
| nsv833957 | Chr.20:32755316 - 32907992 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map