Assay Details
Target Gene Details
Entrez Gene ID: | 4773 |
Gene Name: | nuclear factor of activated T-cells 2 |
Gene Aliases: |
NFAT1, NFATP |
Location: |
Chr.20:51386957-51562857 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFATC2 | NM_001136021.2 | 11 | 7148 | NP_001129493.1 |
| NM_001258292.1 | 10 | 7060 | NP_001245221.1 | |
| NM_001258294.1 | 11 | 6634 | NP_001245223.1 | |
| NM_001258295.1 | 10 | 6546 | NP_001245224.1 | |
| NM_001258296.1 | 11 | 6712 | NP_001245225.1 | |
| NM_001258297.1 | 10 | 6624 | NP_001245226.1 | |
| NM_012340.4 | 11 | 7226 | NP_036472.2 | |
| NM_173091.3 | 10 | 7138 | NP_775114.1 | |
| XM_017027851.1 | 9 | 6439 | XP_016883340.1 | |
| BE674599.1 | 1 | 305 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751917 | Chr.20:51052156 - 51598401 on Build GRCh38 | Gain |
|
| nsv1055542 | Chr.20:51378130 - 51402077 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map