Assay Details
Target Gene Details
Entrez Gene ID: | 149954 |
Gene Name: | BPI fold containing family B member 4 |
Gene Aliases: |
C20orf186, LPLUNC4, RY2G5, dJ726C3.5 |
Location: |
Chr.20:33081512-33111751 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPIFB4 | NM_182519.2 | NP_872325.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
| esv2758792 | Chr.20:32908001 - 33160360 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map