Assay Details
Target Gene Details
Entrez Gene ID: | 57593 |
Gene Name: | early B-cell factor 4 |
Gene Aliases: |
COE4, O/E-4 |
Location: |
Chr.20:2692878-2760108 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBF4 | NM_001110514.1 | NP_001103984.1 | ||
| XM_006723600.1 | XP_006723663.1 | |||
| XM_017027983.1 | XP_016883472.1 | |||
| XM_017027984.1 | XP_016883473.1 | |||
| AB037863.1 | BAA92680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519426 | Chr.20:2687083 - 2701560 on Build GRCh38 | Loss |
|
| nsv1057464 | Chr.20:2650623 - 2989686 on Build GRCh38 | Gain |
|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|
| nsv585275 | Chr.20:2693073 - 2696059 on Build GRCh38 | Loss |
|
| nsv458852 | Chr.20:2657933 - 2704278 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map