Assay Details
Target Gene Details
Entrez Gene ID: | 140893 |
Gene Name: | RBBP8 N-terminal like |
Gene Aliases: |
C20orf151 |
Location: |
Chr.20:62410237-62427573 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBBP8NL | NM_080833.2 | NP_543023.2 | ||
| AK074919.1 | BAC11293.1 | |||
| AK128718.1 | 3 | 2054 | BAC87589.1 | |
| AK290299.1 | ||||
| BC006450.1 | AAH06450.1 | |||
| BC033142.1 | AAH33142.2 | |||
| BC137326.1 | ||||
| BC137327.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422398 | Chr.20:62207833 - 62674040 on Build GRCh38 | Duplication |
|
| nsv586496 | Chr.20:62332390 - 62812195 on Build GRCh38 | Gain |
|
| nsv470557 | Chr.20:62312525 - 62415019 on Build GRCh38 | Gain |
|
| nsv586497 | Chr.20:62345988 - 62415019 on Build GRCh38 | Loss |
|
| nsv953302 | Chr.20:62345345 - 62831248 on Build GRCh38 | Deletion |
|
| nsv518296 | Chr.20:62203245 - 62415019 on Build GRCh38 | Loss |
|
| dgv7651n54 | Chr.20:62315508 - 62427975 on Build GRCh38 | Loss |
|
| nsv586498 | Chr.20:62387568 - 62415019 on Build GRCh38 | Loss |
|
| nsv586500 | Chr.20:62394547 - 62421984 on Build GRCh38 | Gain |
|
| dgv4343n100 | Chr.20:62391262 - 62569333 on Build GRCh38 | Gain |
|
| nsv834024 | Chr.20:62259914 - 62455522 on Build GRCh38 | Gain |
|
| nsv470556 | Chr.20:62212004 - 62577283 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map