Assay Details
Target Gene Details
Entrez Gene ID: | 7032 |
Gene Name: | trefoil factor 2 |
Gene Aliases: |
SML1, SP |
Location: |
Chr.21:42346357-42351099 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFF2 | NM_005423.4 | NP_005414.1 | ||
| AA307808.1 | ||||
| BC032820.1 | AAH32820.1 | |||
| BM817851.1 | ||||
| BM853946.1 | ||||
| X51698.1 | CAA35995.1 |
More Information
Set Membership: |
|

Genomic Map