Assay Details
Target Gene Details
Entrez Gene ID: | 89765 |
Gene Name: | radial spoke head 1 homolog |
Gene Aliases: |
CT79, RSP44, RSPH10A, TSA2, TSGA2 |
Location: |
Chr.21:42472486-42496354 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSPH1 | NM_001286506.1 | NP_001273435.1 | ||
| NM_080860.3 | NP_543136.1 | |||
| XM_005261208.2 | XP_005261265.1 | |||
| XM_011529786.1 | XP_011528088.1 | |||
| AB006536.1 | BAB83694.1 | |||
| AB041016.1 | BAB92995.1 | |||
| AK057315.1 | ||||
| AK314746.1 | ||||
| BC101519.1 | AAI01520.1 | |||
| BC113367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522001 | Chr.21:42421796 - 42827928 on Build GRCh38 | Gain |
|
| nsv834103 | Chr.21:42456611 - 42612638 on Build GRCh38 | Gain |
|
| nsv587633 | Chr.21:42485217 - 42631745 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117231749] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map