Assay Details
Target Gene Details
Entrez Gene ID: | 100133286 |
Gene Name: | uncharacterized LOC100133286 |
Gene Aliases: |
- |
Location: |
Chr.21:36069642-36126640 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100133286 | NR_040084.1 | 4 | 1158 | |
| AK303953.1 | 4 | 1158 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751927 | Chr.21:36061467 - 36286211 on Build GRCh38 | Gain |
|
| nsv587461 | Chr.21:36003345 - 36087866 on Build GRCh38 | Loss |
|
| nsv470899 | Chr.21:36001424 - 36096978 on Build GRCh38 | Loss |
|
| nsv964547 | Chr.21:36003423 - 36090261 on Build GRCh38 | Deletion |
|

Genomic Map