Assay Details
Target Gene Details
Entrez Gene ID: | 3753 |
Gene Name: | potassium voltage-gated channel subfamily E regulatory subunit 1 |
Gene Aliases: |
ISK, JLNS, JLNS2, LQT2/5, LQT5, MinK |
Location: |
Chr.21:34446688-34512275 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNE1 | NM_000219.5 | NP_000210.2 | ||
| NM_001270402.2 | NP_001257331.1 | |||
| NM_001270403.2 | NP_001257332.1 | |||
| NM_001270404.2 | NP_001257333.1 | |||
| AK304336.1 | ||||
| BC046224.1 | AAH46224.1 | |||
| DA418828.1 | ||||
| DB214219.1 | ||||
| DB223354.1 | ||||
| DB230009.1 | ||||
| DB233067.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7824n54 | Chr.21:34352440 - 34535115 on Build GRCh38 | Gain |
|
| dgv4426n100 | Chr.21:34339322 - 34534919 on Build GRCh38 | Gain |
|
| nsv521323 | Chr.21:34384531 - 34515061 on Build GRCh38 | Gain |
|
| nsv1055429 | Chr.21:34299853 - 34516938 on Build GRCh38 | Gain |
|
| esv3575386 | Chr.21:34353927 - 34534570 on Build GRCh38 | Gain |
|
| esv2762109 | Chr.21:34350696 - 34531755 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79761910] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map