Assay Details
Target Gene Details
Entrez Gene ID: | 11200 |
Gene Name: | checkpoint kinase 2 |
Gene Aliases: |
CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1 |
Location: |
Chr.22:28687743-28741866 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9895 | Chr.22:28633973 - 28788289 on Build GRCh38 | Gain+Loss |
|
| nsv1064624 | Chr.22:28669540 - 28734740 on Build GRCh38 | Gain |
|
| nsv588881 | Chr.22:28597195 - 28713048 on Build GRCh38 | Loss |
|
| nsv834169 | Chr.22:28699458 - 28907941 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map