Assay Details
Target Gene Details
Entrez Gene ID: | 114794 |
Gene Name: | extracellular leucine rich repeat and fibronectin type III domain containing 2 |
Gene Aliases: |
LRRC62, PPP1R29 |
Location: |
Chr.22:37340644-37427470 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELFN2 | NR_110512.1 | |||
| AK055475.1 | ||||
| DA365834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834188 | Chr.22:37210658 - 37396179 on Build GRCh38 | Loss |
|
| nsv829203 | Chr.22:36998126 - 37773322 on Build GRCh38 | Loss |
|
| nsv471196 | Chr.22:37296897 - 37397474 on Build GRCh38 | Loss |
|
| nsv834189 | Chr.22:37283448 - 37447346 on Build GRCh38 | Gain+Loss |
|
| nsv834190 | Chr.22:37293005 - 37419461 on Build GRCh38 | Loss |
|
| nsv817992 | Chr.22:37283630 - 37411264 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75120840] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map