Assay Details
Target Gene Details
Entrez Gene ID: | 85358 |
Gene Name: | SH3 and multiple ankyrin repeat domains 3 |
Gene Aliases: |
DEL22q13.3, PROSAP2, PSAP2, SCZD15, SPANK-2 |
Location: |
Chr.22:50674642-50733212 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHANK3 | NM_033517.1 | NP_277052.1 | ||
| AB569469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1067172 | Chr.22:50586195 - 50677098 on Build GRCh38 | Gain |
|
| dgv784n67 | Chr.22:50675884 - 50740070 on Build GRCh38 | Gain |
|
| nsv955204 | Chr.22:50670973 - 50686372 on Build GRCh38 | Deletion |
|
| dgv1338e212 | Chr.22:50665264 - 50677098 on Build GRCh38 | Gain |
|
| nsv459933 | Chr.22:50667128 - 50727236 on Build GRCh38 | Loss |
|
| dgv1341e212 | Chr.22:50668330 - 50677098 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map