Assay Details
Target Gene Details
Entrez Gene ID: | 400931 |
Gene Name: | MIRLET7B host gene |
Gene Aliases: |
linc-Ppara |
Location: |
Chr.22:46085997-46113928 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIRLET7BHG | NR_027033.2 | |||
| NR_110479.1 | ||||
| AK023424.1 | ||||
| AK130875.1 | BAC85451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv459904 | Chr.22:46079585 - 46103554 on Build GRCh38 | Loss |
|
| nsv471212 | Chr.22:46027058 - 46138166 on Build GRCh38 | Loss |
|
| nsv471213 | Chr.22:46079584 - 46119835 on Build GRCh38 | Gain |
|
| nsv829281 | Chr.22:45861485 - 46735858 on Build GRCh38 | Gain |
|
| nsv829285 | Chr.22:46024689 - 46114974 on Build GRCh38 | Gain |
|

Genomic Map