Assay Details
Target Gene Details
Entrez Gene ID: | 6300 |
Gene Name: | mitogen-activated protein kinase 12 |
Gene Aliases: |
ERK-6, ERK3, ERK6, MAPK 12, P38GAMMA, PRKM12, SAPK-3, SAPK3 |
Location: |
Chr.22:50252902-50261759 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK12 | NM_001303252.1 | NP_001290181.1 | ||
| NM_002969.4 | NP_002960.2 | |||
| AK289770.1 | ||||
| AK294422.1 | ||||
| AK307421.1 | ||||
| AK308768.1 | ||||
| BC015741.1 | AAH15741.1 | |||
| BX640648.1 | CAE45794.1 | |||
| CB123561.1 | ||||
| CR456515.1 | CAG30401.1 | |||
| U66243.1 | AAB40118.1 | |||
| X79483.1 | CAA55984.1 | |||
| Y10487.1 | CAA71511.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955199 | Chr.22:50149372 - 50320271 on Build GRCh38 | Deletion |
|
| nsv471223 | Chr.22:50190317 - 50271919 on Build GRCh38 | Loss |
|
| dgv781n67 | Chr.22:50153213 - 50314433 on Build GRCh38 | Gain |
|
| nsv1066496 | Chr.22:50226183 - 50292445 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map