Assay Details
Target Gene Details
Entrez Gene ID: | 646851 |
Gene Name: | family with sequence similarity 227 member A |
Gene Aliases: |
- |
Location: |
Chr.22:38578120-38656629 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM227A | NM_001013647.1 | NP_001013669.1 | ||
| NM_001291030.1 | NP_001277959.1 | |||
| XM_006724291.2 | XP_006724354.2 | |||
| XM_006724292.2 | XP_006724355.2 | |||
| XM_006724293.2 | XP_006724356.1 | |||
| XM_006724294.2 | XP_006724357.2 | |||
| XM_006724295.2 | XP_006724358.2 | |||
| XM_011530308.2 | XP_011528610.1 | |||
| XM_011530309.2 | XP_011528611.1 | |||
| XM_011530310.1 | XP_011528612.1 | |||
| XM_011530311.1 | XP_011528613.1 | |||
| XM_011530312.1 | XP_011528614.1 | |||
| XM_011530313.2 | XP_011528615.1 | |||
| XM_011530314.1 | XP_011528616.1 | |||
| AK301819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3624 | Chr.22:38552181 - 38602777 on Build GRCh38 | Deletion |
|

Genomic Map