Assay Details
Target Gene Details
Entrez Gene ID: | 80115 |
Gene Name: | BAI1 associated protein 2 like 2 |
Gene Aliases: |
- |
Location: |
Chr.22:38084889-38110944 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAIAP2L2 | NM_025045.5 | NP_079321.3 | ||
| XM_005261751.4 | XP_005261808.1 | |||
| XM_011530379.2 | XP_011528681.1 | |||
| XM_011530380.2 | XP_011528682.1 | |||
| XM_011530381.2 | XP_011528683.1 | |||
| XM_011530382.2 | XP_011528684.1 | |||
| XM_011530383.2 | XP_011528685.1 | |||
| XM_011530384.2 | XP_011528686.1 | |||
| XM_011530386.2 | XP_011528688.1 | |||
| XM_011530387.2 | XP_011528689.1 | |||
| XM_011530388.2 | XP_011528690.1 | |||
| AK026235.1 | 9 | 1629 | BAB15405.1 | |
| AY358164.1 | AAQ88531.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471198 | Chr.22:38019714 - 38149612 on Build GRCh38 | Loss |
|
| nsv829205 | Chr.22:38090278 - 38105969 on Build GRCh38 | Loss |
|
| nsv834194 | Chr.22:38063495 - 38240504 on Build GRCh38 | Loss |
|
| nsv834193 | Chr.22:37997329 - 38172090 on Build GRCh38 | Loss |
|
| nsv829206 | Chr.22:38091339 - 38097015 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map