Assay Details
Target Gene Details
Entrez Gene ID: | 84133 |
Gene Name: | zinc and ring finger 3 |
Gene Aliases: |
BK747E2.3, RNF203 |
Location: |
Chr.22:28800657-29057488 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNRF3 | NM_001206998.1 | NP_001193927.1 | ||
| NM_032173.3 | NP_115549.2 | |||
| XM_011530435.2 | XP_011528737.1 | |||
| XM_011530436.2 | XP_011528738.1 | |||
| XM_011530437.1 | XP_011528739.1 | |||
| XM_011530438.2 | XP_011528740.1 | |||
| XM_017028990.1 | XP_016884479.1 | |||
| AB051436.1 | BAB33319.1 | |||
| AK096397.1 | ||||
| CR456397.1 | CAG30283.1 |
Target Gene Details
Entrez Gene ID: | 100874123 |
Gene Name: | ZNRF3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.22:29024999-29031476 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNRF3-AS1 | NR_046851.1 | |||
| AK095488.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064079 | Chr.22:28669540 - 29060071 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116035657] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map