Assay Details
Target Gene Details
Entrez Gene ID: | 4733 |
Gene Name: | developmentally regulated GTP binding protein 1 |
Gene Aliases: |
NEDD3 |
Location: |
Chr.22:31399553-31434186 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DRG1 | NM_004147.3 | NP_004138.1 | ||
| AF078103.1 | AAD12240.1 | |||
| AJ005940.1 | CAA06775.1 | |||
| AK098576.1 | ||||
| AK315659.1 | ||||
| AL133085.1 | CAB61403.1 | |||
| BC019285.1 | AAH19285.1 | |||
| BC020803.1 | AAH20803.1 | |||
| BT007237.1 | AAP35901.1 | |||
| CR456488.1 | CAG30374.1 | |||
| CR542059.1 | CAG46856.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834176 | Chr.22:31286440 - 31468715 on Build GRCh38 | Loss |
|
| nsv1063634 | Chr.22:31401018 - 31525268 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map