Assay Details
Target Gene Details
Entrez Gene ID: | 89781 |
Gene Name: | HPS4, biogenesis of lysosomal organelles complex 3 subunit 2 |
Gene Aliases: |
BLOC3S2, LE |
Location: |
Chr.22:26443613-26483863 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPS4 | XM_011530492.1 | XP_011528794.1 | ||
| XM_017029045.1 | XP_016884534.1 | |||
| XM_017029048.1 | XP_016884537.1 | |||
| XM_017029049.1 | XP_016884538.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs150352517] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map