Assay Details
Target Gene Details
Entrez Gene ID: | 5355 |
Gene Name: | proteolipid protein 2 |
Gene Aliases: |
A4, A4LSB |
Location: |
Chr.X:49171837-49175120 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLP2 | NM_002668.2 | NP_002659.1 | ||
| BC109066.2 | AAI09067.1 | |||
| DB220888.1 | ||||
| L09604.1 | AAA35499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map