Assay Details
Target Gene Details
Entrez Gene ID: | 554 |
Gene Name: | arginine vasopressin receptor 2 |
Gene Aliases: |
ADHR, DI1, DIR, DIR3, NDI, V2R |
Location: |
Chr.X:153902531-153907166 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AVPR2 | NM_000054.4 | NP_000045.1 | ||
| NM_001146151.1 | NP_001139623.1 | |||
| NR_027419.1 | ||||
| XM_006724828.3 | XP_006724891.1 | |||
| AF030626.1 | AAB86428.1 | |||
| AF032388.1 | AAB87678.1 | |||
| AF101727.1 | AAD16444.1 | |||
| AF101728.1 | AAD16445.1 | |||
| AY242131.1 | AAO92298.1 | |||
| BC101484.1 | AAI01485.1 | |||
| BC112181.1 | AAI12182.1 | |||
| BG830436.1 | ||||
| Z11687.1 | CAA77746.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| nsv469867 | Chr.X:153813281 - 154018926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map