Assay Details
Target Gene Details
Entrez Gene ID: | 54440 |
Gene Name: | SAM and SH3 domain containing 3 |
Gene Aliases: |
753P9, CXorf9, HACS2, SH3D6C, SLY |
Location: |
Chr.X:129779916-129795201 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SASH3 | NM_018990.3 | NP_061863.1 | ||
| XM_006724763.1 | XP_006724826.1 | |||
| AK292023.1 | ||||
| AK296665.1 | ||||
| AK314869.1 | ||||
| AL049683.1 | CAB41255.1 | |||
| BC051881.1 | AAH51881.1 | |||
| CR975883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
| nsv526197 | Chr.X:129672435 - 129826254 on Build GRCh38 | Gain |
|
| esv2758893 | Chr.X:129563598 - 129830109 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map