Assay Details
Target Gene Details
Entrez Gene ID: | 158800 |
Gene Name: | Rhox homeobox family member 1 |
Gene Aliases: |
OTEX, PEPP1 |
Location: |
Chr.X:120109049-120117651 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOXF1 | XM_011531281.2 | XP_011529583.1 |
Target Gene Details
Entrez Gene ID: | 101928969 |
Gene Name: | RHOXF1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:120036236-120146855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOXF1-AS1 | NR_131238.1 | |||
| AK123976.1 | ||||
| DA929866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526653 | Chr.X:120029666 - 120140727 on Build GRCh38 | Gain |
|
| esv2758888 | Chr.X:119823476 - 120195131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map