Assay Details
Target Gene Details
Entrez Gene ID: | 158801 |
Gene Name: | NFKB activating protein pseudogene 1 |
Gene Aliases: |
CXorf42 |
Location: |
Chr.X:120236452-120245267 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKAPP1 | NR_027131.1 | |||
| AK093895.1 | BAC04246.1 | |||
| BX537594.1 | 1 | 1749 | CAD97792.1 | |
| DB004517.1 |

Genomic Map