Assay Details
Target Gene Details
Entrez Gene ID: | 6855 |
Gene Name: | synaptophysin |
Gene Aliases: |
MRX96, MRXSYP |
Location: |
Chr.X:49187812-49200202 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYP | NM_003179.2 | NP_003170.1 | ||
| AK295524.1 | ||||
| AK313030.1 | ||||
| AK315953.1 | ||||
| BC064550.1 | AAH64550.1 | |||
| X06389.1 | CAA29686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map