Assay Details
Target Gene Details
Entrez Gene ID: | 23096 |
Gene Name: | IQ motif and Sec7 domain 2 |
Gene Aliases: |
BRAG1, MRX1, MRX78 |
Location: |
Chr.X:53225784-53321350 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQSEC2 | XM_006724582.3 | 16 | 5075 | XP_006724645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map