Assay Details
Target Gene Details
Entrez Gene ID: | 3920 |
Gene Name: | lysosomal associated membrane protein 2 |
Gene Aliases: |
CD107b, LAMP-2, LAMPB, LGP110 |
Location: |
Chr.X:120426148-120469349 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMP2 | NM_001122606.1 | 9 | 2582 | NP_001116078.1 |
| NM_002294.2 | 9 | 5418 | NP_002285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528786 | Chr.X:120380837 - 120471759 on Build GRCh38 | Gain |
|
| esv2758889 | Chr.X:120267570 - 120449094 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map