Assay Details
Target Gene Details
Entrez Gene ID: | 100129520 |
Gene Name: | TEX13 family member C |
Gene Aliases: |
- |
Location: |
Chr.X:125319871-125325214 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEX13C | NM_001195272.1 | 1 | 2920 | NP_001182201.1 |
| XM_005262352.3 | 1 | 3169 | XP_005262409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752314 | Chr.X:125319728 - 125808304 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map