Assay Details
Target Gene Details
Entrez Gene ID: | 100873921 |
Gene Name: | SYP antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:49198839-49202454 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYP-AS1 | NR_046649.1 | |||
| AI018666.1 | ||||
| BX104027.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|

Genomic Map