Assay Details
Target Gene Details
Entrez Gene ID: | 7569 |
Gene Name: | zinc finger protein 182 |
Gene Aliases: |
HHZ150, KOX14, ZNF21, Zfp182 |
Location: |
Chr.X:47974851-48003995 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF182 | NM_001007088.1 | NP_001007089.1 | ||
| NM_001178099.1 | NP_001171570.1 | |||
| NM_006962.1 | NP_008893.1 | |||
| AK122874.1 | ||||
| AK299325.1 | ||||
| BC106875.2 | AAI06876.1 | |||
| BC114216.1 | AAI14217.1 | |||
| CD642892.1 | ||||
| CN311292.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522886 | Chr.X:47978461 - 48062292 on Build GRCh38 | Gain |
|
| esv2758866 | Chr.X:47907511 - 48287275 on Build GRCh38 | Gain+Loss |
|
| dgv17n29 | Chr.X:47976492 - 48213935 on Build GRCh38 | Gain+Loss |
|
| nsv6890 | Chr.X:47968293 - 48168190 on Build GRCh38 | Deletion |
|
| nsv471601 | Chr.X:47976493 - 48213935 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73491288] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map