Assay Details
Target Gene Details
Entrez Gene ID: | 3920 |
Gene Name: | lysosomal associated membrane protein 2 |
Gene Aliases: |
CD107b, LAMP-2, LAMPB, LGP110 |
Location: |
Chr.X:120426148-120469349 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMP2 | NM_001122606.1 | NP_001116078.1 | ||
| NM_002294.2 | NP_002285.1 | |||
| AK291090.1 | ||||
| AK295038.1 | ||||
| AY561849.1 | AAS67876.1 | |||
| J04183.1 | AAA60383.1 | |||
| X77196.1 | CAA54416.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528786 | Chr.X:120380837 - 120471759 on Build GRCh38 | Gain |
|
| esv2758889 | Chr.X:120267570 - 120449094 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146840553] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map