Assay Details
Target Gene Details
Entrez Gene ID: | 645864 |
Gene Name: | MAGE family member B17 |
Gene Aliases: |
- |
Location: |
Chr.X:16167481-16172394 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB17 | NM_001277307.1 | NP_001264236.1 | ||
| XM_011545567.2 | XP_011543869.1 | |||
| DV080535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436917 | Chr.X:16046179 - 16176281 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map