Assay Details
Target Gene Details
Entrez Gene ID: | 51213 |
Gene Name: | leucine zipper protein 4 |
Gene Aliases: |
CT-28, CT-8, CT28, HOM-TES-85 |
Location: |
Chr.X:115289717-115307556 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUZP4 | NM_001318840.1 | NP_001305769.1 | ||
| NM_016383.4 | NP_057467.1 | |||
| AF124430.1 | AAF28870.1 | |||
| AK093369.1 | ||||
| AK314765.1 | ||||
| BC128134.1 | ||||
| BC128135.1 | ||||
| DB035084.1 | ||||
| DB041564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577051 | Chr.X:115223253 - 115839825 on Build GRCh38 | Gain |
|
| nsv520351 | Chr.X:115073935 - 115318424 on Build GRCh38 | Gain |
|
| esv33994 | Chr.X:113685854 - 115738426 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map