Assay Details
Target Gene Details
Entrez Gene ID: | 347541 |
Gene Name: | MAGE family member B5 |
Gene Aliases: |
CT3.3, MAGE-B5 |
Location: |
Chr.X:26216169-26218270 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB5 | NM_001271752.1 | NP_001258681.1 | ||
| CR747535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522405 | Chr.X:26208070 - 27089369 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map